DL-Propargylglycine 98+%

Supplier: Ambeed
Warning

A1473787-100G A1473787-1G A1473787-25G A1473787-5G A1473787-250MG
76663-122EA 1803 USD
76663-122 76663-124 76663-126 76663-128 77875-105
DL-Propargylglycine 98+%
DL-Propargylglycine

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MDL Number: MFCD00056728
CAS Number: 64165-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR