N-Methylmorpholine-N-oxide ~50% (w/w) in aqueous solution

Supplier: Ambeed
Warning

A1473779-100G A1473779-25G A1473779-500G
76663-116EA 20 USD
76663-116 76663-118 76663-120
N-Methylmorpholine-N-oxide ~50% (w/w) in aqueous solution
N-Methylmorpholine-N-oxide

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00005947
CAS Number: 7529-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR