4-Oxo-4H-pyran-2,6-dicarboxylic acid 98% + (contains < 2% H₂O)

Supplier: Ambeed
Warning

Synonyms: Chelidonic acid, 2,6-Dicarboxy-4-oxo-4H-pyran

A1473713-1G A1473713-25G A1473713-5G
76663-104EA 23.34 USD
76663-104 76663-106 76663-108
4-Oxo-4H-pyran-2,6-dicarboxylic acid 98% + (contains < 2% H₂O)
4-Oxo-4H-pyran-2,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄O₆
MW: 184.103 g/mol
MDL Number: MFCD00006577
CAS Number: 99-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR