3-Methyl-1-(piperidin-4-yl)azetidin-3-ol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1473651-100MG
A1473651-1G
A1473651-250MG
76663-092EA
153
USD
76663-092
76663-094
76663-096
3-Methyl-1-(piperidin-4-yl)azetidin-3-ol hydrochloride 95%
3-Methyl-1-(piperidin-4-yl)azetidin-3-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₉ClN₂O |
CAS Number:
2493424-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...