2-Amino-4-methoxybenzothiazole 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

A1473514-100G A1473514-1G A1473514-25G A1473514-5G
76663-082EA 581.38 USD
76663-082 76663-084 76663-086 76663-088
2-Amino-4-methoxybenzothiazole 98+%
2-Amino-4-methoxybenzothiazole

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00005792
CAS Number: 5464-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR