2-Methoxycinnamic acid 98+% predominantly trans

Supplier: Ambeed
Warning

Synonyms: 3-(2-Methoxyphenyl)acrylic acid, 2-Methoxycinnamic Acid

A1473496-100G A1473496-1G A1473496-25G A1473496-500G A1473496-5G
76663-072EA 40.32 USD
76663-072 76663-074 76663-076 76663-078 76663-080
2-Methoxycinnamic acid 98+% predominantly trans
2-Methoxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD00064238
CAS Number: 6099-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR