4-Methylhex-1-yn-3-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1473494-100MG
A1473494-1G
A1473494-250MG
76663-066EA
340.55
USD
76663-066
76663-068
76663-070
4-Methylhex-1-yn-3-amine hydrochloride 95%
4-Methylhex-1-yn-3-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClN MW: 147.6458 g/mol |
MDL Number:
MFCD34186975 CAS Number: 2703752-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...