1,3-Bis((R)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Danger
|
A1473490-100MG
A1473490-1G
A1473490-250MG
76663-064EA
51.99
USD
76663-064
77624-260
77624-262
1,3-Bis((R)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate 97%
1,3-Bis((R)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₁H₂₅BF₄N₂ MW: 392.2412 g/mol |
CAS Number:
2757082-93-0 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...