1,3-Bis((S)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1473488-100MG A1473488-250MG
76663-062EA 57.29 USD
76663-062 77597-440
1,3-Bis((S)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate 97%
1,3-Bis((S)-1-phenylpropyl)-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₅BF₄N₂
MW: 392.2412 g/mol
CAS Number: 569658-95-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR