1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium chloride 97%

Supplier: Ambeed
Warning

A1473482-100MG A1473482-1G A1473482-250MG
76663-050EA 62.6 USD
76663-050 76663-052 76663-054
1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium chloride 97%
1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₁₉H₂₁ClN₂
MW: 312.8364 g/mol
CAS Number: 186354-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR