1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1473480-100MG A1473480-250MG A1473480-1G A1473480-5G
76663-048EA 64 USD
76663-048 77597-438 77622-226 77875-099
1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate 97%
1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₉H₂₁BF₄N₂
CAS Number: 613686-48-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR