1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Danger
|
A1473478-100MG
A1473478-250MG
76663-044EA
59
USD
76663-044
76663-046
1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate 97%
1,3-Bis((R)-1-phenylethyl)-1H-imidazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₉H₂₁BF₄N₂ |
CAS Number:
569658-93-1 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...