1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride 97%

Supplier: Ambeed
Warning

A1473472-100MG A1473472-1G A1473472-250MG
76663-038EA 102 USD
76663-038 76663-040 76663-042
1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride 97%
1,3-Bis((S)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₂₇H₂₅ClN₂
CAS Number: 188840-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR