1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride 97%
Supplier: Ambeed
|
Danger
|
A1473470-100MG
A1473470-1G
A1473470-250MG
76663-036EA
102
USD
76663-036
77597-428
77597-430
1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride 97%
1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₂₇H₂₅ClN₂ |
CAS Number:
186354-48-3 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...