9-(2-Bromophenyl)phenanthrene 98%

Supplier: Ambeed
Warning

A1473318-1G A1473318-250MG A1473318-5G
76663-030EA 43 USD
76663-030 76663-032 76663-034
9-(2-Bromophenyl)phenanthrene 98%
9-(2-Bromophenyl)phenanthrene

Precaución: for Research Use Only

Formula: C₂₀H₁₃Br
MDL Number: MFCD32695537
CAS Number: 3582-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR