3-Hydroxy-4-methyl-benzaldehyde ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-methylbenzaldehyde

A1473125-1G A1473125-5G
76663-008EA 81.69 USD
76663-008 76663-010
3-Hydroxy-4-methyl-benzaldehyde ≥98%
3-Hydroxy-4-methyl-benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.148 g/mol
MDL Number: MFCD01632496
CAS Number: 57295-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR