6-Bromo-2,3-difluorotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4-difluoro-2-methylbenzene

A1473016-1G A1473016-25G A1473016-5G
76663-002EA 18.04 USD
76663-002 76663-004 76663-006
6-Bromo-2,3-difluorotoluene 95%
6-Bromo-2,3-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD07777161
CAS Number: 847502-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR