2,5-Dimethoxybenzene-1,4-diamine dihydrochloride 98%

Supplier: Ambeed
Warning

A1472944-100MG A1472944-250MG
76662-992EA 44 USD
76662-992 76662-994
2,5-Dimethoxybenzene-1,4-diamine dihydrochloride 98%
2,5-Dimethoxybenzene-1,4-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂N₂O₂
CAS Number: 25445-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR