3-Bromo-1H-indazole 98+%

Supplier: Ambeed
Danger

A1472822-1G A1472822-25G A1472822-5G
76662-980EA 17 USD
76662-980 76662-982 76662-984
3-Bromo-1H-indazole 98+%
3-Bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD00159926
CAS Number: 40598-94-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR