(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride 97% (by HPLC)

Supplier: Ambeed
Warning

A1472715-100G A1472715-1G A1472715-25G A1472715-5G
76662-966EA 81.69 USD
76662-966 76662-968 76662-970 76662-972
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride 97% (by HPLC)
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₁₉ClN₂O₄
MW: 386.82 g/mol
MDL Number: MFCD09842269
CAS Number: 171752-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR