Ibudilast 99%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Isopropylpyrazolo[1,5-a]pyridin-3-yl)-2-methylpropan-1-one, 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine

A1472713-100MG A1472713-1G A1472713-250MG A1472713-25G A1472713-5G
76662-956EA 28.65 USD
76662-956 76662-958 76662-960 76662-962 76662-964
Ibudilast 99%
Ibudilast

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O
MW: 230.31 g/mol
MDL Number: MFCD00864808
CAS Number: 50847-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR