Pyridin-2-amine hydrochloride 95% (contains < 30% H₂O)

Supplier: Ambeed
Warning

A1472661-250MG A1472661-5G
76662-938EA 13 USD
76662-938 76662-940
Pyridin-2-amine hydrochloride 95% (contains < 30% H₂O)
Pyridin-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂
MDL Number: MFCD00137029
CAS Number: 32654-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR