N-(2-Hydroxyethyl)morpholine 99%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Hydroxyethyl)morpholine, 2-Morpholinoethanol

A1472547-100G A1472547-25G A1472547-500G
76662-930EA 10 USD
76662-930 76662-932 76662-934
N-(2-Hydroxyethyl)morpholine 99%
N-(2-Hydroxyethyl)morpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00006180
CAS Number: 622-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR