3-Fluoro-5-(trifluoromethoxy)aniline hydrochloride 95%

Supplier: Ambeed
Warning

A1471754-100MG A1471754-1G A1471754-250MG
76662-924EA 61 USD
76662-924 76662-926 76662-928
3-Fluoro-5-(trifluoromethoxy)aniline hydrochloride 95%
3-Fluoro-5-(trifluoromethoxy)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₇H₆ClF₄NO
MDL Number: MFCD34186971
CAS Number: 2703756-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR