1,2,3,4-Tetrahydroisoquinoline 98%

Supplier: Ambeed
Warning

A1471640-25G A1471640-500G A1471640-5G
76662-914EA 20 USD
76662-914 76662-916 76662-918
1,2,3,4-Tetrahydroisoquinoline 98%
1,2,3,4-Tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₁N
MDL Number: MFCD00006896
CAS Number: 91-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR