3,5-Dimethyl-4-hydroxybenzonitrile 99+%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile

A1471604-100G A1471604-25G A1471604-5G
76662-908EA 46 USD
76662-908 76662-910 76662-912
3,5-Dimethyl-4-hydroxybenzonitrile 99+%
3,5-Dimethyl-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00221716
CAS Number: 4198-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR