6,6'-Dimethyl-2,2'-bipyridyl 99%

Supplier: Ambeed
Warning

Synonyms: 6,6'-Dimethyl-2,2'-bipyridine

A1471590-100G A1471590-1G A1471590-25G A1471590-5G
76662-898EA 564.4 USD
76662-898 76662-900 76662-902 76662-904
6,6'-Dimethyl-2,2'-bipyridyl 99%
6,6'-Dimethyl-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.2371 g/mol
MDL Number: MFCD00059776
CAS Number: 4411-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR