1-(2-Methoxyphenyl)-2-phenoxyethanol 95%

Supplier: Ambeed
Warning

A1471568-100MG A1471568-250MG
76662-894EA 44.56 USD
76662-894 76662-896
1-(2-Methoxyphenyl)-2-phenoxyethanol 95%
1-(2-Methoxyphenyl)-2-phenoxyethanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃
MW: 244.2857 g/mol
CAS Number: 1273871-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR