(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 99% +99%ee

Supplier: Ambeed
Danger

Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

A1471307-1G A1471307-25G A1471307-5G A1471307-100G
76662-876EA 6 USD
76662-876 76662-878 76662-880 77597-408
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 99% +99%ee
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
MDL Number: MFCD00671527
CAS Number: 68737-65-5
UN: 3259
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR