trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥98%, ee 99%

Supplier: Ambeed
Danger

Synonyms: (1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, (1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine

A1471293-5G A1471293-1G A1471293-25G
76662-874EA 18.04 USD
76662-874 77597-404 77597-406
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥98%, ee 99%
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
MDL Number: MFCD00671528
CAS Number: 87583-89-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR