(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine 99+%

Supplier: Ambeed
Warning

A1471259-100G A1471259-1G A1471259-25G A1471259-5G
76662-866EA 171 USD
76662-866 76662-868 76662-870 76662-872
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine 99+%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine

Precaución: for Research Use Only

Formula: C₃₆H₅₄N₂O₂
MDL Number: MFCD00191800
CAS Number: 135616-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR