(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride 99+%
Supplier: Ambeed
|
Warning
|
A1471257-100G
A1471257-1G
A1471257-25G
A1471257-5G
76662-858EA
195
USD
76662-858
76662-860
76662-862
76662-864
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride 99+%
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride
Precaución: for Research Use Only
Formula:
C₃₆H₅₄Cl₃MnN₂O₂ |
MDL Number:
MFCD02101663 CAS Number: 135620-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...