(1R,2R)-rel-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine 98+%

Supplier: Ambeed
Warning

A1471253-100G A1471253-25G A1471253-5G
76662-846EA 492.26 USD
76662-846 76662-848 76662-850
(1R,2R)-rel-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine 98+%
(1R,2R)-rel-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂
MW: 170.29 g/mol
MDL Number: MFCD31689292
CAS Number: 43148-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR