trans-N1,N1-Dimethylcyclohexane-1,2-diamine 98%

Supplier: Ambeed
Warning

A1471249-100G A1471249-1G A1471249-25G A1471249-5G
76662-830EA 523 USD
76662-830 76662-832 76662-834 76662-836
trans-N1,N1-Dimethylcyclohexane-1,2-diamine 98%
trans-N1,N1-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MDL Number: MFCD09838678
CAS Number: 67198-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR