Ferrocenemethanol ≥98%
Supplier: Ambeed
|
Warning
|
A1471247-100G
A1471247-1G
A1471247-5G
A1471247-25G
76662-824EA
479
USD
76662-824
76662-826
76662-828
77875-073
Ferrocenemethanol ≥98%
Ferrocenemethanol
Precaución: for Research Use Only
Formula:
C₁₁H₁₂FeO |
MDL Number:
MFCD00040878 CAS Number: 1273-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...