(±)-trans-1,2-Diaminocyclohexane 98%

Supplier: Ambeed
Danger

Synonyms: (±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine

A1471237-100G A1471237-5G
76662-816EA 79.57 USD
76662-816 76662-818
(±)-trans-1,2-Diaminocyclohexane 98%
(±)-trans-1,2-Diaminocyclohexane

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00063747
CAS Number: 1121-22-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR