(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 99%

Supplier: Ambeed
Warning

A1471235-100G A1471235-25G A1471235-500G A1471235-5G
76662-808EA 71 USD
76662-808 76662-810 76662-812 76662-814
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 99%
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00191979
CAS Number: 39961-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR