(1S,2S)-(+)-1,2-Diaminocyclohexane ≥98%, ee 99%
Supplier: Ambeed
|
Danger
|
A1471233-1G
A1471233-25G
A1471233-5G
76662-802EA
10
USD
76662-802
76662-804
76662-806
(1S,2S)-(+)-1,2-Diaminocyclohexane ≥98%, ee 99%
(1S,2S)-(+)-1,2-Diaminocyclohexane
Precaución: for Research Use Only
Formula:
C₆H₁₄N₂ |
MDL Number:
MFCD00062986 CAS Number: 21436-03-3 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...