(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate 99%

Supplier: Ambeed
Warning

A1471229-100G A1471229-25G A1471229-500G A1471229-5G
76662-790EA 66 USD
76662-790 76662-792 76662-794 76662-796
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate 99%
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00191980
CAS Number: 67333-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR