1-Bromo-5-chloropentane ≥99%
Supplier: Ambeed
|
Warning
|
A1471072-100G
A1471072-25G
A1471072-500G
76662-782EA
16
USD
76662-782
76662-784
76662-786
1-Bromo-5-chloropentane ≥99%
1-Bromo-5-chloropentane
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₃CH₂Cl |
MDL Number:
MFCD00001016 CAS Number: 54512-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...