1,10-Dichlorodecane 99+%

Supplier: Ambeed
Danger

A1471070-100G A1471070-25G A1471070-500G
76662-776EA 15 USD
76662-776 76662-778 76662-780
1,10-Dichlorodecane 99+%
1,10-Dichlorodecane

Precaución: for Research Use Only

Formula: ClH₂C(CH₂)₈CH₂Cl
MDL Number: MFCD00000957
CAS Number: 2162-98-3
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR