1-Bromododecane 99%

Supplier: Ambeed
Warning

A1471062-1000G A1471062-100G A1471062-25G A1471062-500G
76662-762EA 50 USD
76662-762 76662-764 76662-766 76662-768
1-Bromododecane 99%
1-Bromododecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₁₀CH₃
MDL Number: MFCD00000225
CAS Number: 143-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR