1-Bromodecane 99%

Supplier: Ambeed
Danger

Synonyms: 1-Decyl bromide

A1471060-25G A1471060-100G A1471060-500G
76662-760EA 6.37 USD
76662-760 77567-024 77622-210
1-Bromodecane 99%
1-Bromodecane

Precaución: For research use only.

Formula: BrH₂C(CH₂)₈CH₃
MW: 221.18 g/mol
MDL Number: MFCD00000221
CAS Number: 112-29-8
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR