5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine 97%
Supplier: Ambeed
|
Warning
|
A1471050-100MG
A1471050-1G
A1471050-250MG
76662-748EA
93.36
USD
76662-748
76662-750
76662-752
5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine 97%
5-(11bR)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine
Precaución: for Research Use Only
Formula:
C₃₄H₂₄NO₂P MW: 509.5336 g/mol |
CAS Number:
2093047-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...