5-(11bS)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine 97%
Supplier: Ambeed
|
Warning
|
A1471048-100MG
A1471048-1G
A1471048-250MG
76662-742EA
157.02
USD
76662-742
76662-744
76662-746
5-(11bS)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine 97%
5-(11bS)-Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl-10,11-dihydro-5H-dibenz[b,f]azepine
Precaución: for Research Use Only
Formula:
C₃₄H₂₄NO₂P MW: 509.5336 g/mol |
CAS Number:
1361055-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...