2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole 99% (by HPLC)

Supplier: Ambeed
Warning

A1471018-100G A1471018-25G A1471018-500G
76662-736EA 68 USD
76662-736 76662-738 76662-740
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole 99% (by HPLC)
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole

Precaución: for Research Use Only

Formula: C₄₂H₂₈Cl₂N₄
MDL Number: MFCD00060081
CAS Number: 1707-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR