3-(tert-Butyl)-5-(dimethylamino)-2-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A1470638-100MG A1470638-1G A1470638-250MG A1470638-5G
76662-708EA 27.59 USD
76662-708 76662-710 76662-712 76662-714
3-(tert-Butyl)-5-(dimethylamino)-2-hydroxybenzaldehyde 95%
3-(tert-Butyl)-5-(dimethylamino)-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.2955 g/mol
CAS Number: 335596-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR