1,3-Dimesityl-4,5-dimethyl-1H-imidazol-3-ium chloride 95%

Supplier: Ambeed
Warning

A1470566-10G A1470566-1G A1470566-25G A1470566-5G
76662-692EA 514.54 USD
76662-692 76662-694 76662-696 76662-698
1,3-Dimesityl-4,5-dimethyl-1H-imidazol-3-ium chloride 95%
1,3-Dimesityl-4,5-dimethyl-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₂₃H₂₉ClN₂
MW: 368.94 g/mol
CAS Number: 1118916-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR