(7,9-Bis(2,6-diisopropylphenyl)-7,9-dihydro-8H-acenaphtho[1,2-d]imidazol-8-ylidene)gold(III) chloride 98%
Supplier: Ambeed
|
Warning
|
A1470415-100MG
A1470415-1G
A1470415-250MG
76662-686EA
79.57
USD
76662-686
76662-688
76662-690
(7,9-Bis(2,6-diisopropylphenyl)-7,9-dihydro-8H-acenaphtho[1,2-d]imidazol-8-ylidene)gold(III) chloride 98%
(7,9-Bis(2,6-diisopropylphenyl)-7,9-dihydro-8H-acenaphtho[1,2-d]imidazol-8-ylidene)gold(III) chloride
Precaución: for Research Use Only
Formula:
C₃₇H₄₀AuClN₂ MW: 745.1465 g/mol |
CAS Number:
1246183-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...