9H-Carbazole-2,7-dicarboxylic acid 95% (by HPLC) (contains < 10% DMF)
Supplier: Ambeed
|
Warning
|
A1469863-100MG
A1469863-250MG
76662-682EA
45.62
USD
76662-682
76662-684
9H-Carbazole-2,7-dicarboxylic acid 95% (by HPLC) (contains < 10% DMF)
9H-Carbazole-2,7-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₉NO₄ MW: 255.23 g/mol |
CAS Number:
1409971-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...